Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-Dimethylacryloyl chloride

Catalog No. 18366025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-366-025G 25 g
18-366-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-366-025G Supplier Sigma Aldrich Supplier No. 18366025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,3-dimethylacryloyl chloride reacts with silylketene acetals in acetonitrile to give α-ethylenic α-keto esters.3,3-dimethylacryloyl chloride was used in the synthesis of:
  • Substituted 4,4-dimethyl-3,4-dihydro-1h-quinolin-2-one via condensation with aniline
  • N-(3,3-dimethylacryloyl)-(s)-leucine methyl ester

Specifications

CAS 3350-78-5
Density 1.065 g/mL (at 25°C (literature))
Boiling Point 145°C to 147°C (lit.)
Molecular Formula C5H7ClO
Refractive Index n20/D 1.476 (literature)
Linear Formula (CH3)2 C=CHCOCl
MDL Number MFCD00000730
Quantity 25 g
Synonym 3-Methyl-2-butenoyl chloride; 3-Methylcrotonoyl chloride; Senecioyl chloride
Molecular Weight (g/mol) 118.56
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.