Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethylacryloyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3350-78-5
C5H7ClO
118.56
MFCD00000730
BDUBTLFQHNYXPC-UHFFFAOYSA-N
3,3-dimethylacryloyl chloride,3-methylcrotonoyl chloride,3-methyl-but-2-enoyl chloride,3,3-dimethylacrylyl chloride,2-butenoyl chloride, 3-methyl,3-methyl-2-butenoyl chloride,3,3-dimethylacryloylchloride,dimethylacrylyl chloride,dimethyl acryloyl chloride,acmc-1cpb0
102394
3-methylbut-2-enoyl chloride
CC(=CC(=O)Cl)C
3350-78-5
C5H7ClO
118.56
MFCD00000730
BDUBTLFQHNYXPC-UHFFFAOYSA-N
3,3-dimethylacryloyl chloride,3-methylcrotonoyl chloride,3-methyl-but-2-enoyl chloride,3,3-dimethylacrylyl chloride,2-butenoyl chloride, 3-methyl,3-methyl-2-butenoyl chloride,3,3-dimethylacryloylchloride,dimethylacrylyl chloride,dimethyl acryloyl chloride,acmc-1cpb0
102394
3-methylbut-2-enoyl chloride
CC(=CC(=O)Cl)C
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 51°C
Specific Gravity 1.06
Quantity 25g
Refractive Index 1.4750 to 1.4780
Color Colorless to Yellow
Boiling Point 145.0°C to 147.0°C
Density 1.0600g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2C=CHCOCl
Beilstein 02, 433
Solubility Information Solubility in water: reacts
Formula Weight 118.56
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3, 3-Dimethylacryloyl chloride, 97%
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 222-109-4

TSCA : TSCA

Recommended Storage : Product may darken on storage