Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-Dimethylallyl bromide

Catalog No. 24990425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-990-425G 25 g
24-990-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-990-425G Supplier Sigma Aldrich Supplier No. 24990425G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,3-dimethylallyl bromide has been used in the preparation of:
  • (+/-)-eldanolide
  • 1-(3,3-dimethylallyl)-L-tryptophan
  • (+/-)-fumagillin, antibiotic isolated from aspergillus fumigattus

Specifications

CAS 870-63-3
Density 1.29 g/mL (at 20°C (literature))
Molecular Formula C5H9Br
Linear Formula (CH3)2 C=CHCH2Br
MDL Number MFCD00000242
Boiling Point 82°C to 83°C (150 mmHg)
Synonym 1-Bromo-3-methyl-2-butene; Prenyl bromide
Molecular Weight (g/mol) 149.03
Quantity 25 g
Percent Purity ∼90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.