Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3-Dimethylallyl bromide

Catalog No. 2743721G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-437-21G 1 g
27-437-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-437-21G Supplier Sigma Aldrich Supplier No. 2743721G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,3-dimethylallyl bromide was used in the synthesis of 1- and 2-allylic-3-methylindoles
  • It was also used in the synthesis of products and base-modified pyrimidine nucleosides

Specifications

CAS 870-63-3
Density 1.29 g/mL (at 20°C (literature))
Boiling Point 82°C to 83°C (150 mmHg)
Molecular Formula C5H9Br
Refractive Index n20/D 1.489 (literature)
Linear Formula (CH3)2 C=CHCH2Br
MDL Number MFCD00000242
Quantity 1 g
Synonym 1-Bromo-3-methyl-2-butene; Prenyl bromide
Molecular Weight (g/mol) 149.03
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.