Learn More
eMolecules​ 3,3-DIMETHYLBUTANAL5G
Supplier:  eMolecules​ 572143404
Encompass_Preferred
Ambeed | 3,3-Dimethylbutanal | 5g | A187800 | Purity: % | 2987-16-8 | MFCD00042807 | MW: 100.16
Catalog No. NC2293775