Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenylpropionic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
606-83-7
C15H14O2
226.27
MFCD00002717
BZQGAPWJKAYCHR-UHFFFAOYSA-N
3,3-diphenylpropionic acid,benzenepropanoic acid, .beta.-phenyl,3,3-diphenyl-propionic acid,propionic acid, 3,3-diphenyl,.beta.-phenylbenzenepropanoic acid,beta,beta-diphenylpropionic acid,hydrocinnamic acid, .beta.-phenyl,beta-phenylbenzenepropanoic acid,hydrocinnamic acid, beta-phenyl,pubchem13618
64798
3,3-diphenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)C2=CC=CC=C2
606-83-7
C15H14O2
226.27
MFCD00002717
BZQGAPWJKAYCHR-UHFFFAOYSA-N
3,3-diphenylpropionic acid,benzenepropanoic acid, .beta.-phenyl,3,3-diphenyl-propionic acid,propionic acid, 3,3-diphenyl,.beta.-phenylbenzenepropanoic acid,beta,beta-diphenylpropionic acid,hydrocinnamic acid, .beta.-phenyl,beta-phenylbenzenepropanoic acid,hydrocinnamic acid, beta-phenyl,pubchem13618
64798
3,3-diphenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)C2=CC=CC=C2
Flash Point 113°C
Packaging Glass bottle
Quantity 25g
Color White to Yellow
Melting Point 150°C to 155°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (C6H5)2CHCH2CO2H
Beilstein 09, 680
Solubility Information Solubility in water: slightly soluble
Formula Weight 226.27
Physical Form Crystalline Powder
Percent Purity 96.0 to 104.0% Acid, ≥96.0% (GC)
Chemical Name or Material 3, 3-Diphenylpropionic acid, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-125-4

TSCA : TSCA