Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenylpropylamine 98.0+%, TCI America™
SDP

Catalog No. D185525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D185525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D185525G Supplier TCI America Supplier No. D185525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5586-73-2
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD00008202
InChI Key KISZTEOELCMZPY-UHFFFAOYSA-N
Synonym 3,3-diphenylpropylamine, benzenepropanamine, .gamma.-phenyl, 3-3-diphenylpropylamine, 3,3-diphenylpropan-1-amin, acmc-209lpx, 3.3-diphenylpropylamine, 3,3 diphenylpropylamine, 3,3-diphenyl propylamine, 3,3-diphenyl-propylamine, 3,3-diphenylpropylamine e
PubChem CID 79698
IUPAC Name 3,3-diphenylpropan-1-amine
SMILES C1=CC=C(C=C1)C(CCN)C2=CC=CC=C2

Specifications

Color White-Yellow
Boiling Point 166°C
Quantity 25 g
Formula Weight 211.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 3,3-Diphenylpropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.