Learn More
Cayman Chemical 3,3′-Dipropylthiadicarbocyanine (iodide), Purity: ≥98%, CAS: 53213-94-8, 50 mg
SDP
Catalog No. 502912961
Encompass_Preferred
A fluorescent dye; has been used to monitor cell membrane potential; ex/em = 622/670 or 622/688 nm when cells are depolarized or hyperpolarized, respectively; fluorescence intensity decreases in hyperpolarized cells and increases in depolarized cells; induces membrane hyperpolarization, increases in intracellular sodium, and decreases in intracellular potassium in Ehrlich ascites tumor cells at 3 µM; increases potassium membrane permeability by 64.6% in Ehrlich ascites tumor cells in the presence of ouabain; reduces cellular ATP levels in the absence of glucose and induces acid production and inhibits oxygen consumption in the presence of glucose in Ehrlich ascites tumor cells at 3 µM
Catalog No. 50-291-2961 Supplier Cayman Chemical Supplier No. 3630850 MG
A fluorescent dye; has been used to monitor cell membrane potential; ex/em = 622/670 or 622/688 nm when cells are depolarized or hyperpolarized, respectively; fluorescence intensity decreases in hyperpolarized cells and increases in depolarized cells; induces membrane hyperpolarization, increases in intracellular sodium, and decreases in intracellular potassium in Ehrlich ascites tumor cells at 3 µM; increases potassium membrane permeability by 64.6% in Ehrlich ascites tumor cells in the presence of ouabain; reduces cellular ATP levels in the absence of glucose and induces acid production and inhibits oxygen consumption in the presence of glucose in Ehrlich ascites tumor cells at 3 µM