Learn More
Medchemexpress LLC 3,3'-dithiobispropionic acid bis-sulfosuccinimidyl ester disodium salt | 81069-02-5 | MFCD00054980 | 98.9% | 564.54 g/mol | C14H16N2O14S4 | 5 MG
SDP
Catalog No. 5000202215
Encompass_Preferred
DTSSP (3,3'-dithiobis(sulfosuccinimidyl propionate)) is a cleavable disulfide-containing crosslinker used in biochemical research as a reversible protein crosslinker and as a linker in antibody-drug conjugate (ADC) synthesis. It couples to primary amines via NHS-ester chemistry to form stable amide bonds and is designed to be cleaved under reducing conditions to release linked moieties.
  • Cleavable disulfide-containing crosslinker enabling reversible linkage.
  • Reacts with primary amines via NHS-ester chemistry to form stable amide bonds.
  • Cleavable under reducing conditions for controlled release of conjugates.
  • Suitable for antibody-drug conjugate linker synthesis and protein crosslinking.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-02215 Supplier Medchemexpress LLC Supplier No. HY1263495MG
DTSSP (3,3'-dithiobis(sulfosuccinimidyl propionate)) is a cleavable disulfide-containing crosslinker used in biochemical research as a reversible protein crosslinker and as a linker in antibody-drug conjugate (ADC) synthesis. It couples to primary amines via NHS-ester chemistry to form stable amide bonds and is designed to be cleaved under reducing conditions to release linked moieties.
  • Cleavable disulfide-containing crosslinker enabling reversible linkage.
  • Reacts with primary amines via NHS-ester chemistry to form stable amide bonds.
  • Cleavable under reducing conditions for controlled release of conjugates.
  • Suitable for antibody-drug conjugate linker synthesis and protein crosslinking.
  • Available in small laboratory pack sizes for research use.