Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3'-DithiodIpr∗

Catalog No. D3669500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3669500MG 1 g
D36691G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. D3669500MG Supplier Sigma Aldrich Supplier No. D3669500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,3'sa-DithiodIpr∗

Specifications

CAS 57757-57-0
Molecular Formula C14H16N2O8S2
Linear Formula C14H16N2O8S2
MDL Number MFCD00042045
Quantity 1 g
Synonym DTSP; Di(N-succinimidyl) 3,3'-dithiodipropionate; Dithiobis(succinimidyl propionate); Lomant's reagent
Molecular Weight (g/mol) 404.42 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.