Learn More
Medchemexpress LLC 3,3'-dithiodipropionic acid | 1119-62-6 | MFCD00002780 | 99.0% | 210.26 | C6H10O4S2 | 50 G
SDP
Catalog No. 5000302126
Encompass_Preferred
Dithiodipropionic acid is a disulfide-containing dicarboxylic acid used as a research reagent in chemical and biological studies. It is supplied as a solid material for use in synthesis, formulation studies, and the design of redox-responsive constructs, and is reported to form nanoaggregates with certain small molecules.
  • High purity (99.0%) suitable for research applications.
  • Disulfide-containing dicarboxylic structure for redox-responsive design.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 210.26 and formula C6H10O4S2 for formulation calculations.
  • Compatible with DMSO for dissolution and stock solutions.
Catalog No. 50-003-02126 Supplier Medchemexpress LLC Supplier No. HYW01439550G
Dithiodipropionic acid is a disulfide-containing dicarboxylic acid used as a research reagent in chemical and biological studies. It is supplied as a solid material for use in synthesis, formulation studies, and the design of redox-responsive constructs, and is reported to form nanoaggregates with certain small molecules.
  • High purity (99.0%) suitable for research applications.
  • Disulfide-containing dicarboxylic structure for redox-responsive design.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 210.26 and formula C6H10O4S2 for formulation calculations.
  • Compatible with DMSO for dissolution and stock solutions.