Learn More
Medchemexpress LLC 3,3'-dithiodipropionic acid | 1119-62-6 | MFCD00002780 | 99.0% | 210.26 g/mol | C6H10O4S2 | 1 ML
SDP
Catalog No. 5000303938
Encompass_Preferred
Dithiodipropionic acid is a sulfur-containing dicarboxylic acid supplied for laboratory research. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid quantities. The compound has molecular formula C6H10O4S2, molecular weight 210.26 g/mol, and a reported purity of 99.0%. Supporting documents including a data sheet, certificate of analysis, and SDS are available, and storage recommendations are provided for both powder and solvent solutions.
  • Ready-to-use 10 mM solution in DMSO for immediate experiments.
  • High purity (99.0%) suitable for analytical and research use.
  • Available as solid in multiple gram sizes for synthesis and scale-up.
  • Highly soluble in DMSO (≥100 mg/mL); use newly opened DMSO due to hygroscopicity.
  • Supporting documents provided: data sheet, COA, and SDS.
  • Storage guidance included for powder and in-solution conditions.
Catalog No. 50-003-03938 Supplier Medchemexpress LLC Supplier No. HYW01439510MM/1ML
Dithiodipropionic acid is a sulfur-containing dicarboxylic acid supplied for laboratory research. It is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid quantities. The compound has molecular formula C6H10O4S2, molecular weight 210.26 g/mol, and a reported purity of 99.0%. Supporting documents including a data sheet, certificate of analysis, and SDS are available, and storage recommendations are provided for both powder and solvent solutions.
  • Ready-to-use 10 mM solution in DMSO for immediate experiments.
  • High purity (99.0%) suitable for analytical and research use.
  • Available as solid in multiple gram sizes for synthesis and scale-up.
  • Highly soluble in DMSO (≥100 mg/mL); use newly opened DMSO due to hygroscopicity.
  • Supporting documents provided: data sheet, COA, and SDS.
  • Storage guidance included for powder and in-solution conditions.