Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Fluorophenyl)propionic Acid 97.0+%, TCI America™
SDP

Catalog No. F07791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07791G 1 g
F07795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07791G Supplier TCI America Supplier No. F07791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 458-45-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD01310821
InChI Key UBLMRADOKLXLCD-UHFFFAOYSA-N
Synonym 3-3-fluorophenyl propionic acid, 3-3-fluorophenyl propanoic acid, 3-fluorohydrocinnamic acid, 3-3-fluoro-phenyl-propionic acid, 3-fluoro-benzenepropanoic acid, benzenepropanoic acid, 3-fluoro, pubchem13947, acmc-209k4f, 3-fluorobenzenepropanoic acid, benzenepropanoic acid,3-fluoro
PubChem CID 2737489
IUPAC Name 3-(3-fluorophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)F)CCC(=O)O

Specifications

Melting Point 43°C
Boiling Point 119°C
Quantity 1 g
Formula Weight 168.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3-Fluorophenyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.