Learn More
Medchemexpress LLC 3'-((3-hydroxy-2,4-dioxo-3,4-dihydrothieno[3,2-d]pyrimidin-1(2H)-yl)methyl)-biphenyl-4-carboxamide | 824983-94-0 | 99.9% | 25 MG
SDP
Catalog No. 5000231008
Encompass_Preferred
FEN1-IN-2 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) used for biochemical and cellular research. It potently inhibits FEN1 (IC50 3 nM) and shows activity against XPG (IC50 226 nM).
  • Potent FEN1 inhibitor with an IC50 of 3 nM.
  • Shows measurable activity against XPG (IC50 226 nM).
  • High purity suitable for research use (99.94%).
  • Molecular weight 393.42 g/mol, formula C20H15N3O4S.
  • Available in 25 MG quantities for laboratory experiments.
Catalog No. 50-002-31008 Supplier Medchemexpress LLC Supplier No. HY12269525MG
FEN1-IN-2 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) used for biochemical and cellular research. It potently inhibits FEN1 (IC50 3 nM) and shows activity against XPG (IC50 226 nM).
  • Potent FEN1 inhibitor with an IC50 of 3 nM.
  • Shows measurable activity against XPG (IC50 226 nM).
  • High purity suitable for research use (99.94%).
  • Molecular weight 393.42 g/mol, formula C20H15N3O4S.
  • Available in 25 MG quantities for laboratory experiments.