Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Hydroxyphenyl)-3-hydroxypropionic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002168 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2168 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2168 Supplier MilliporeSigma Supelco Supplier No. 0670410MG
Add to Cart
Add to Cart

Specifications

CAS 3247-75-4
Linear Formula C9H10O4
MDL Number MFCD16749143
Synonym 3-(3-Hydroxyphenyl)-3-hydroxypropanoic acid; 3-(3-Hydroxyphenyl)hydracrylic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 182.17
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.