Learn More
Medchemexpress LLC 3-(3-Hydroxyphenyl)propionic acid | 621-54-5 | 166.18 | 10 MM 1 ML
SDP
Catalog No. 5000449064
Encompass_Preferred
3-(3-Hydroxyphenyl)propionic acid (3HPPA) is an endothelium-dependent nitric oxide (NO) release promoter and endothelial nitric oxide synthase (eNOS) activator. It activates eNOS to mediate vascular smooth muscle relaxation and enhances endothelial cell NO synthesis, inducing vasodilation and reducing peripheral vascular resistance. It has antihypertensive and vascular protective effects and can be used in the prevention and treatment of cardiovascular diseases.
  • Activates eNOS to promote NO release
  • Mediates vascular smooth muscle relaxation
  • Enhances endothelial cell NO synthesis
  • Induces vasodilation and reduces peripheral vascular resistance
  • Exhibits antihypertensive and vascular protective effects
Catalog No. 50-004-49064 Supplier Medchemexpress LLC Supplier No. HYW00525510MM/1ML
3-(3-Hydroxyphenyl)propionic acid (3HPPA) is an endothelium-dependent nitric oxide (NO) release promoter and endothelial nitric oxide synthase (eNOS) activator. It activates eNOS to mediate vascular smooth muscle relaxation and enhances endothelial cell NO synthesis, inducing vasodilation and reducing peripheral vascular resistance. It has antihypertensive and vascular protective effects and can be used in the prevention and treatment of cardiovascular diseases.
  • Activates eNOS to promote NO release
  • Mediates vascular smooth muscle relaxation
  • Enhances endothelial cell NO synthesis
  • Induces vasodilation and reduces peripheral vascular resistance
  • Exhibits antihypertensive and vascular protective effects