Learn More
Medchemexpress LLC 3-(3-methoxyphenyl)propionic acid | 10516-71-9 | MFCD00014027 | 98.0% | 180.20 g/mol | C10H12O3 | 1 ML
SDP
Catalog No. 5000300777
Encompass_Preferred
3-(3-methoxyphenyl)propionic acid is an organic acid and an endogenous human metabolite excreted in urine. Supplied for laboratory research in solution and solid formats, the compound is provided with supporting documentation for safe handling and analytical use.
  • Naturally occurring human metabolite.
  • Available as 10 mM solution in DMSO and in solid pack sizes.
  • High purity suitable for research (98.03%).
  • Molecular formula C10H12O3; molecular weight 180.20 g/mol.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-00777 Supplier Medchemexpress LLC Supplier No. HYW01648210MM/1ML
3-(3-methoxyphenyl)propionic acid is an organic acid and an endogenous human metabolite excreted in urine. Supplied for laboratory research in solution and solid formats, the compound is provided with supporting documentation for safe handling and analytical use.
  • Naturally occurring human metabolite.
  • Available as 10 mM solution in DMSO and in solid pack sizes.
  • High purity suitable for research (98.03%).
  • Molecular formula C10H12O3; molecular weight 180.20 g/mol.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
  • For research use only; not for human or clinical use.