Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3-methyl-3H-diazirin-3-yl)propanoic acid | 25055-86-1 | MFCD19228230 | >97.0% | 128.13 g/mol | C5H8N2O2 | 250 MG
SDP

Catalog No. 5000302466
Encompass_Preferred

3-(3-methyl-3H-diazirin-3-yl)propanoic acid is a diazirine-containing carboxylic acid used as a chemical intermediate and photoreactive building block in organic and medicinal chemistry. It is employed to introduce a photo-crosslinking diazirine moiety into target molecules for probe synthesis and study of biomolecular interactions.

  • Small, 128.13 g/mol carboxylic acid with formula C5H8N2O2.
  • Contains a diazirine photoreactive group for carbene generation upon irradiation.
  • Useful as a synthetic intermediate for probe and drug analog synthesis.
  • Enables photoaffinity labeling and covalent capture of interaction partners.
  • Typically supplied at high purity suitable for research applications (suppliers list ≥97%).

Catalog No. 50-003-02466 Supplier Medchemexpress LLC Supplier No. 250MGHYW024615
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(3-methyl-3H-diazirin-3-yl)propanoic acid is a diazirine-containing carboxylic acid used as a chemical intermediate and photoreactive building block in organic and medicinal chemistry. It is employed to introduce a photo-crosslinking diazirine moiety into target molecules for probe synthesis and study of biomolecular interactions.

  • Small, 128.13 g/mol carboxylic acid with formula C5H8N2O2.
  • Contains a diazirine photoreactive group for carbene generation upon irradiation.
  • Useful as a synthetic intermediate for probe and drug analog synthesis.
  • Enables photoaffinity labeling and covalent capture of interaction partners.
  • Typically supplied at high purity suitable for research applications (suppliers list ≥97%).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.