Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium Bromide] 98.0+%, TCI America™
SDP

Catalog No. M27915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27915G 5 g
M27911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27915G Supplier TCI America Supplier No. M27915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 889617-36-1
Molecular Formula C31H42Br2N4
Molecular Weight (g/mol) 630.513
InChI Key ZZGWEIDPDBDPOH-UHFFFAOYSA-L
PubChem CID 78198750
IUPAC Name 1-[2,6-di(propan-2-yl)phenyl]-3-[[3-[2,6-di(propan-2-yl)phenyl]imidazol-1-ium-1-yl]methyl]imidazol-3-ium;dibromide
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2)C[N+]3=CN(C=C3)C4=C(C=CC=C4C(C)C)C(C)C.[Br-].[Br-]

Specifications

Melting Point 277°C
Color White
Quantity 5 g
Formula Weight 630.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium Bromide]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.