Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Methylenediindole 98.0+%, TCI America™
SDP

Catalog No. M13995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13995G 5 g
M139925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13995G Supplier TCI America Supplier No. M13995G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1968-05-4
Molecular Formula C17H14N2
Molecular Weight (g/mol) 246.31
MDL Number MFCD00195766
InChI Key VFTRKSBEFQDZKX-UHFFFAOYSA-N
Synonym 3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole
PubChem CID 3071
ChEBI CHEBI:50182
IUPAC Name 3-(1H-indol-3-ylmethyl)-1H-indole
SMILES C1=CC=C2C(=C1)C(=CN2)CC3=CNC4=CC=CC=C43

Specifications

Melting Point 167°C
Quantity 5 g
Formula Weight 246.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Methylenediindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.