Learn More
Medchemexpress LLC 3 3 - Propane-2 2-di 5g
SDP
Catalog No. 5000205680
Encompass_Preferred
3 3 -(Propane-2 2-Diylbis (sulfanediyl))dipropionic acid is a ROS-sensitive cleavable linker that can be used in the synthesis of ADCs 3 3 -(Propane-2 2-diylbis(sulfanediyl))dipropionic acid is promising for research of tumor drug delivery systems[1]
Catalog No. 50-002-05680 Supplier Medchemexpress LLC Supplier No. HYW1092245G
3 3 -(Propane-2 2-Diylbis (sulfanediyl))dipropionic acid is a ROS-sensitive cleavable linker that can be used in the synthesis of ADCs 3 3 -(Propane-2 2-diylbis(sulfanediyl))dipropionic acid is promising for research of tumor drug delivery systems[1]