Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3-Trifluoromethylphenyl)propanol | 78573-45-2 | C10H11F3O | 25 G
SDP

Catalog No. 5000357621
Encompass_Preferred

3-(3-Trifluoromethylphenyl)propanol is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It also finds application as a laboratory chemical and in the manufacture of other substances. This compound appears as a liquid with a boiling point of 236.9°C at 760 mmHg and a flash point of 104.7±17.0°C. Its relative density is approximately 1.203±0.06 g/cm3.

  • Drug intermediate for synthesis of active compounds
  • Suitable for laboratory research and manufacturing of substances
  • Boiling point: 236.9°C at 760 mmHg
  • Flash point: 104.7±17.0°C
  • Relative density: 1.203±0.06 g/cm3
  • Store at 4°C, sealed and away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-57621 Supplier Medchemexpress LLC Supplier No. 25GHY76039
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(3-Trifluoromethylphenyl)propanol is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It also finds application as a laboratory chemical and in the manufacture of other substances. This compound appears as a liquid with a boiling point of 236.9°C at 760 mmHg and a flash point of 104.7±17.0°C. Its relative density is approximately 1.203±0.06 g/cm3.

  • Drug intermediate for synthesis of active compounds
  • Suitable for laboratory research and manufacturing of substances
  • Boiling point: 236.9°C at 760 mmHg
  • Flash point: 104.7±17.0°C
  • Relative density: 1.203±0.06 g/cm3
  • Store at 4°C, sealed and away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.