Learn More
Cayman Chemical 3 3DIthIodIpropIonIc ACD d 1g
SDP
Catalog No. 502947747
Encompass_Preferred
A protein cross-linking reagent; has been used as a molecular ruler to determine protein tertiary and quaternary structure; has been used as a component of self-assembled monolayers in gold-based biosensors
Catalog No. 50-294-7747 Supplier Cayman Chemical Supplier No. 334051 G
A protein cross-linking reagent; has been used as a molecular ruler to determine protein tertiary and quaternary structure; has been used as a component of self-assembled monolayers in gold-based biosensors