Learn More
Cayman Chemical 3,3′-Dithiodipropionic Acid di(N-hydroxysuccinimide ester), Purity: ≥90%, CAS: 57757-57-0, 250 mg
SDP
Catalog No. 502946990
Encompass_Preferred
A protein cross-linking reagent; has been used as a molecular ruler to determine protein tertiary and quaternary structure; has been used as a component of self-assembled monolayers in gold-based biosensors
Catalog No. 50-294-6990 Supplier Cayman Chemical Supplier No. 33405250 MG
A protein cross-linking reagent; has been used as a molecular ruler to determine protein tertiary and quaternary structure; has been used as a component of self-assembled monolayers in gold-based biosensors