Learn More
Cayman Chemical 3-(3-Hydroxyphenyl)propionic Acid sulfate, Purity: ≥95%, CAS: 86321-32-6, 10 mg
SDP
Catalog No. 502892842
Encompass_Preferred
A metabolite of certain phenols and glycosides; urinary excretion is reduced in C. parvum-infected protein-deficient mice and increased in mice fed wheat bran or functionalized wheat bran
Catalog No. 50-289-2842 Supplier Cayman Chemical Supplier No. 900410310 MG
A metabolite of certain phenols and glycosides; urinary excretion is reduced in C. parvum-infected protein-deficient mice and increased in mice fed wheat bran or functionalized wheat bran