Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[[(3S)-3-(aminomethyl)-1-hydroxy-3H-2,1-benzoxaborol-7-yl]oxy]propan-1-ol hydrochloride | 1234563-16-6 | MFCD28411360 | 99.6% | 273.52 g/mol | C11H17BClNO4 | 25MG
SDP

Catalog No. 5000235126
Encompass_Preferred

Epetraborole hydrochloride is the hydrochloride salt of epetraborole, a boron-containing leucyl-tRNA synthetase inhibitor used in antibacterial research. It inhibits protein synthesis in Gram-negative bacteria and is provided as a solid research reagent for biochemical and microbiological studies.

  • High purity suitable for research (99.6%).
  • Leucyl-tRNA synthetase inhibitor; reported IC50 = 0.31 μM.
  • White to off-white solid, stable under standard storage conditions.
  • Molecular formula C11H17BClNO4; molecular weight 273.52 g/mol.
  • CAS number 1234563-16-6.
  • Supplied in small research quantities for assay development and mechanistic studies.

Catalog No. 50-002-35126 Supplier Medchemexpress LLC Supplier No. HY12479A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epetraborole hydrochloride is the hydrochloride salt of epetraborole, a boron-containing leucyl-tRNA synthetase inhibitor used in antibacterial research. It inhibits protein synthesis in Gram-negative bacteria and is provided as a solid research reagent for biochemical and microbiological studies.

  • High purity suitable for research (99.6%).
  • Leucyl-tRNA synthetase inhibitor; reported IC50 = 0.31 μM.
  • White to off-white solid, stable under standard storage conditions.
  • Molecular formula C11H17BClNO4; molecular weight 273.52 g/mol.
  • CAS number 1234563-16-6.
  • Supplied in small research quantities for assay development and mechanistic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.