Learn More
Cayman Chemical 3,3'-Sulfinyldipropanoic Acid, Purity: ≥98%, CAS: 3680-08-8, 100 mg
SDP
Catalog No. 502915377
Encompass_Preferred
An APE1/Ref-1 inhibitor (IC50 = 40 µM); a building block in the synthesis of DSSO
Catalog No. 50-291-5377 Supplier Cayman Chemical Supplier No. 37214100 MG
An APE1/Ref-1 inhibitor (IC50 = 40 µM); a building block in the synthesis of DSSO