Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [3-[[4-(1,1-dimethylethoxy)phenoxy]methyl]-3,4-dihydro-2(1H)-isoquinolinyl]-1-naphthalenylmethanone | 1380548-02-6 | 99.2% | 465.58 g/mol | C31H31NO3 | 10 MG
SDP

Catalog No. 5000228023
Encompass_Preferred

SR10067 is a potent, selective, brain-penetrant REV-ERB agonist used in research on metabolic and neuropsychiatric disorders. It exhibits high affinity for Rev-Erbβ and Rev-Erbα with reported IC50 values of 160 nM and 170 nM, respectively, and is supplied as a high-purity research-grade small molecule.

  • Potent Rev-Erbα/β agonist (IC50 170 nM and 160 nM).
  • Brain-penetrant, suitable for in vivo research applications.
  • High purity (approximately 99.2%) for reproducible results.
  • Available in multiple pack sizes, including 10 mg.
  • Molecular weight 465.58 g/mol; formula C31H31NO3.
  • CAS 1380548-02-6 for unambiguous identification.

Catalog No. 50-002-28023 Supplier Medchemexpress LLC Supplier No. HY11751610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SR10067 is a potent, selective, brain-penetrant REV-ERB agonist used in research on metabolic and neuropsychiatric disorders. It exhibits high affinity for Rev-Erbβ and Rev-Erbα with reported IC50 values of 160 nM and 170 nM, respectively, and is supplied as a high-purity research-grade small molecule.

  • Potent Rev-Erbα/β agonist (IC50 170 nM and 160 nM).
  • Brain-penetrant, suitable for in vivo research applications.
  • High purity (approximately 99.2%) for reproducible results.
  • Available in multiple pack sizes, including 10 mg.
  • Molecular weight 465.58 g/mol; formula C31H31NO3.
  • CAS 1380548-02-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.