Learn More
Ace Glass, Inc. PTFE insert, 3/4" NPT, 19/38 outer
SDP
Catalog No. 50983468
Encompass
3/4 NPT PTFE insert with 19/38 outer joint,for use with ACE reaction heads.
Catalog No. 50-983-468 Supplier Ace Glass, Inc. Supplier No. 1286608
3/4 NPT PTFE insert with 19/38 outer joint,for use with ACE reaction heads.