Learn More
MWI VETERINARY 3/4" 22-GA MONOJECT NEEDLES
SDP
Catalog No. NC0779669
Encompass_Preferred
Monoject 250 Hypodermic Needle, 22 gauge, 3/4 in. length, color-blue, regular bevel. There are 100 needles per box.
This item is not returnable. View return policy
Catalog No. NC0779669 Supplier MWI VETERINARY Supplier No. 028562
Monoject 250 Hypodermic Needle, 22 gauge, 3/4 in. length, color-blue, regular bevel. There are 100 needles per box.