Learn More
Ace Glass, Inc. PTFE insert, 3/4" NPT, 29/42 outer
SDP
Catalog No. 50977892
Encompass
3/4 NPT PTFE insert with 29/42 outer joint,for use with ACE reaction heads.
Catalog No. 50-977-892 Supplier Ace Glass, Inc. Supplier No. 1286623
3/4 NPT PTFE insert with 29/42 outer joint,for use with ACE reaction heads.