Learn More
Medchemexpress LLC 3-[4-[(2S,5S)-5-(4-chlorobenzyl)-2-methylmorpholin-4-yl]piperidin-1-yl]-1H-1,2,4-triazol-5-amine | 2088453-21-6 | 99.9% | 390.9 g·mol⁻1 | C19H27ClN6O | 5 MG
SDP
Catalog No. 5000223437
Encompass_Preferred
OATD-01 is a first-in-class, orally active small-molecule inhibitor of chitinase enzymes (CHIT1 and AMCase) with low-nanomolar potency and demonstrated antifibrotic efficacy in preclinical models. Developed for treatment of fibrotic and inflammatory pulmonary diseases, it has progressed into clinical studies and is supplied for research use in biochemical, cellular, and in vivo pharmacology. CAS 2088453-21-6; molecular formula C19H27ClN6O.
  • Inhibits CHIT1 and AMCase with low-nanomolar potency.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • Demonstrated antifibrotic activity in multiple preclinical models.
  • High-purity material provided for research applications.
  • Useful for pharmacokinetic and pharmacodynamic investigations.
Catalog No. 50-002-23437 Supplier Medchemexpress LLC Supplier No. HY137464A5MG
OATD-01 is a first-in-class, orally active small-molecule inhibitor of chitinase enzymes (CHIT1 and AMCase) with low-nanomolar potency and demonstrated antifibrotic efficacy in preclinical models. Developed for treatment of fibrotic and inflammatory pulmonary diseases, it has progressed into clinical studies and is supplied for research use in biochemical, cellular, and in vivo pharmacology. CAS 2088453-21-6; molecular formula C19H27ClN6O.
  • Inhibits CHIT1 and AMCase with low-nanomolar potency.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • Demonstrated antifibrotic activity in multiple preclinical models.
  • High-purity material provided for research applications.
  • Useful for pharmacokinetic and pharmacodynamic investigations.