Learn More
Medchemexpress LLC 3-[4-[(3S)-4-(4-Br-benzoyl)-3-(4-Cl-phenyl)piperazine-1-carbonyl]-5-methylpyrazol-1-yl]benzonitrile | 2095486-67-0 | 100.0% | 588.88 g/mol | C29H23BrClN5O2 | 100 MG
SDP
Catalog No. 5000248561
Encompass_Preferred
eIF4A3-IN-1 is a small-molecule inhibitor of the eukaryotic initiation factor eIF4A3 used as a research tool in cellular and animal studies. It selectively binds a non-ATP site on eIF4A3 and has reported antitumor and analgesic activities; supplied as a high-purity solid suitable for in vitro and in vivo applications.
  • Selective eIF4A3 inhibition
  • High purity (99.96%)
  • Available in multiple sizes, including 100 mg
  • High solubility in DMSO (200 mg/mL) for in vitro use
  • Formulation guidance for in vivo dosing (10% DMSO, 90% corn oil)
  • Stable with long-term powder storage at -20 °C
Catalog No. 50-002-48561 Supplier Medchemexpress LLC Supplier No. HY101513100MG
eIF4A3-IN-1 is a small-molecule inhibitor of the eukaryotic initiation factor eIF4A3 used as a research tool in cellular and animal studies. It selectively binds a non-ATP site on eIF4A3 and has reported antitumor and analgesic activities; supplied as a high-purity solid suitable for in vitro and in vivo applications.
  • Selective eIF4A3 inhibition
  • High purity (99.96%)
  • Available in multiple sizes, including 100 mg
  • High solubility in DMSO (200 mg/mL) for in vitro use
  • Formulation guidance for in vivo dosing (10% DMSO, 90% corn oil)
  • Stable with long-term powder storage at -20 °C