Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[4-[4-(4-bromobenzoyl)-3-(4-chlorophenyl)piperazine-1-carbonyl]-5-methylpyrazol-1-yl]benzonitrile | 2095486-67-0 | 100.0% | 588.88 g/mol | C29H23BrClN5O2 | 25 MG
SDP

Catalog No. 5000246509
Encompass_Preferred

eIF4A3-IN-1 is a selective small-molecule inhibitor of the eukaryotic initiation factor 4A3 (eIF4A3) intended for research use. It inhibits nonsense-mediated RNA decay (NMD), binds the non-ATP site of eIF4A3, and has reported anti-tumor and analgesic activities. Chemical identity: C29H23BrClN5O2, MW 588.88 g/mol, CAS 2095486-67-0; purity 99.96%.

  • selective eIF4A3 inhibition (IC50 0.26 μM; Kd 0.043 μM).
  • high reported purity of 99.96%.
  • soluble in DMSO at 200 mg/mL (requires sonication) for in vitro use.
  • formulation compatible with in vivo vehicle of 10% DMSO/90% corn oil at ≥5 mg/mL.
  • powder and solution storage recommendations for compound stability.
  • available in multiple solid package sizes, including a 25 mg option.

Catalog No. 50-002-46509 Supplier Medchemexpress LLC Supplier No. HY10151325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

eIF4A3-IN-1 is a selective small-molecule inhibitor of the eukaryotic initiation factor 4A3 (eIF4A3) intended for research use. It inhibits nonsense-mediated RNA decay (NMD), binds the non-ATP site of eIF4A3, and has reported anti-tumor and analgesic activities. Chemical identity: C29H23BrClN5O2, MW 588.88 g/mol, CAS 2095486-67-0; purity 99.96%.

  • selective eIF4A3 inhibition (IC50 0.26 μM; Kd 0.043 μM).
  • high reported purity of 99.96%.
  • soluble in DMSO at 200 mg/mL (requires sonication) for in vitro use.
  • formulation compatible with in vivo vehicle of 10% DMSO/90% corn oil at ≥5 mg/mL.
  • powder and solution storage recommendations for compound stability.
  • available in multiple solid package sizes, including a 25 mg option.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.