Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,5-naphthyridine

Catalog No. ADE0008611G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0008611G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0008611G Supplier Sigma Aldrich Supplier No. ADE0008611G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,5-naphthyridine

Specifications

Molecular Formula C14H17BN2O2
Linear Formula C14H17BN2O2
MDL Number MFCD13182107
Quantity 10 g
Molecular Weight (g/mol) 256.11
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.