Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[4-(4-hydroxyphenyl)piperazin-1-yl]-1-phenylpyrrolidine-2,5-dione | 522649-59-8 | 99.7% | 351.4 g/mol | C20H21N3O3 | 10MM 1ML
SDP

Catalog No. 5000238829
Encompass_Preferred

Malic enzyme inhibitor ME1 is a small-molecule research reagent that inhibits cytosolic malic enzyme 1 (ME1) with a reported IC50 of 0.15 μM. The compound is supplied in ready-to-use solution formats for in vitro assays and is provided with supporting documentation for safe handling and quality verification.

  • Potent ME1 inhibitor (IC50 0.15 μM).
  • Supplied as 10 mM solution in DMSO, ready for in vitro use.
  • High purity (≈99.7%) with certificate of analysis available.
  • Soluble in DMSO up to 100 mg/mL; warming can improve solubility.
  • Storage stability information provided for powder and solution formats.
  • Documentation includes SDS, COA, and product data sheet for handling and quality details.

Catalog No. 50-002-38829 Supplier Medchemexpress LLC Supplier No. HY12486110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Malic enzyme inhibitor ME1 is a small-molecule research reagent that inhibits cytosolic malic enzyme 1 (ME1) with a reported IC50 of 0.15 μM. The compound is supplied in ready-to-use solution formats for in vitro assays and is provided with supporting documentation for safe handling and quality verification.

  • Potent ME1 inhibitor (IC50 0.15 μM).
  • Supplied as 10 mM solution in DMSO, ready for in vitro use.
  • High purity (≈99.7%) with certificate of analysis available.
  • Soluble in DMSO up to 100 mg/mL; warming can improve solubility.
  • Storage stability information provided for powder and solution formats.
  • Documentation includes SDS, COA, and product data sheet for handling and quality details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.