Learn More
Medchemexpress LLC 3-[4-(4-hydroxyphenyl)piperazin-1-yl]-1-phenylpyrrolidine-2,5-dione | 522649-59-8 | 99.7% | 351.4 g/mol | C20H21N3O3 | 25MG
SDP
Catalog No. 5000235713
Encompass_Preferred
Malic enzyme inhibitor ME1 is a small-molecule probe that selectively inhibits the cytosolic NADP+-dependent malic enzyme 1 (ME1) with an in vitro IC50 of 0.15 μM. Supplied as a high-purity solid, it is intended for use in biochemical and cell-based studies to interrogate ME1-dependent metabolic pathways and related cellular phenotypes.
  • Potent ME1 inhibition (IC50 0.15 μM).
  • High purity (≈99.7%).
  • Available in multiple vial sizes for scalability.
  • Suitable for biochemical and cell-based assays.
  • Supports mechanistic studies of metabolism and disease.
Catalog No. 50-002-35713 Supplier Medchemexpress LLC Supplier No. HY12486125MG
Malic enzyme inhibitor ME1 is a small-molecule probe that selectively inhibits the cytosolic NADP+-dependent malic enzyme 1 (ME1) with an in vitro IC50 of 0.15 μM. Supplied as a high-purity solid, it is intended for use in biochemical and cell-based studies to interrogate ME1-dependent metabolic pathways and related cellular phenotypes.
  • Potent ME1 inhibition (IC50 0.15 μM).
  • High purity (≈99.7%).
  • Available in multiple vial sizes for scalability.
  • Suitable for biochemical and cell-based assays.
  • Supports mechanistic studies of metabolism and disease.