Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4,4′-Trichlorocarbanilide-13C6 | 1216457-76-9 | MFCD00013254 | 99.2% | 321.54 | C7(13C6)H9Cl3N2O | 1 MG
SDP

Catalog No. 5000285027
Encompass_Preferred

Triclocarban-13C6 is a 13C6 isotopically labeled analytical reference standard of 3,4,4′-trichlorocarbanilide intended for use as an internal standard in mass spectrometry and quantitative analysis.

  • Isotopically labeled (13C6) analogue for use as an internal standard.
  • High purity: 99.2%.
  • Molecular weight 321.54 g/mol.
  • Chemical formula C7(13C6)H9Cl3N2O.
  • White to off-white solid physical form.
  • Supplied quantity 1 mg.
  • Storage recommendations: room temperature for solid; in solvent store at -80 °C (2 years) or -20 °C (1 year).

Catalog No. 50-002-85027 Supplier Medchemexpress LLC Supplier No. HYW5859561MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triclocarban-13C6 is a 13C6 isotopically labeled analytical reference standard of 3,4,4′-trichlorocarbanilide intended for use as an internal standard in mass spectrometry and quantitative analysis.

  • Isotopically labeled (13C6) analogue for use as an internal standard.
  • High purity: 99.2%.
  • Molecular weight 321.54 g/mol.
  • Chemical formula C7(13C6)H9Cl3N2O.
  • White to off-white solid physical form.
  • Supplied quantity 1 mg.
  • Storage recommendations: room temperature for solid; in solvent store at -80 °C (2 years) or -20 °C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.