Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5,6-Tetrabromo-o-cresol 96.0+%, TCI America™
SDP

Catalog No. T152825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T152825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T152825G Supplier TCI America Supplier No. T152825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 576-55-6
Molecular Formula C7H4Br4O
Molecular Weight (g/mol) 423.724
MDL Number MFCD00002148
InChI Key GGIDUULRWQOXLR-UHFFFAOYSA-N
Synonym 2,3,4,5-Tetrabromo-6-methylphenol
PubChem CID 11336
IUPAC Name 2,3,4,5-tetrabromo-6-methylphenol
SMILES CC1=C(C(=C(C(=C1Br)Br)Br)Br)O

Specifications

Melting Point 210°C
Color White
Quantity 25 g
Formula Weight 423.72
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5,6-Tetrabromo-o-cresol
RTECSNumber GP3135000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.