Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,4,5,6-Tetrachloro-1,2-benzoquinone

Catalog No. T66015G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T66015G 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T66015G Supplier Sigma Aldrich Supplier No. T66015G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tritolyl phosphate was used as a mediator in the preparation of uranium-sensitive electrode based on membrane containing uranium di-(4-octylphenyl)phosphate as sensor.

Specifications

CAS 2435-53-2
Melting Point 126°C to 129°C (lit.)
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
MDL Number MFCD00001646
Quantity 500 mL
Synonym o-Chloranil
Molecular Weight (g/mol) 245.88
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.