Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5,6-Tetrafluoro-N-methylphthalimide 95.0+%, TCI America™
SDP

Catalog No. T24215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24215G 5 g
T242125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24215G Supplier TCI America Supplier No. T24215G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33795-85-6
Molecular Formula C9H3F4NO2
Molecular Weight (g/mol) 233.12
MDL Number MFCD01631699
InChI Key AULXVJOTNZUFIY-UHFFFAOYSA-N
PubChem CID 469561
IUPAC Name 4,5,6,7-tetrafluoro-2-methyl-2,3-dihydro-1H-isoindole-1,3-dione
SMILES CN1C(=O)C2=C(C1=O)C(F)=C(F)C(F)=C2F

Specifications

Melting Point 160°C
Color Yellow
Quantity 5 g
Formula Weight 233.12
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5,6-Tetrafluoro-N-methylphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.