Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5,6-Tetrafluorosalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. T19881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19881G Supplier TCI America Supplier No. T19881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14742-36-0
Molecular Formula C7H2F4O3
Molecular Weight (g/mol) 210.08
MDL Number MFCD00483242
InChI Key WKYGDMSOKGXAAY-UHFFFAOYSA-N
Synonym 3,4,5,6-Tetrafluoro-2-hydroxybenzoic Acid
PubChem CID 11020133
IUPAC Name 2,3,4,5-tetrafluoro-6-hydroxybenzoic acid
SMILES OC(=O)C1=C(O)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 210.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5,6-Tetrafluorosalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.