Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5,6-Tetrahydrophthalic anhydride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2426-02-0
C8H8O3
152.15
HMMBJOWWRLZEMI-UHFFFAOYSA-N
3,4,5,6-tetrahydrophthalic anhydride,4,5,6,7-tetrahydroisobenzofuran-1,3-dione,1-cyclohexene-1,2-dicarboxylic anhydride,1,3-isobenzofurandione, 4,5,6,7-tetrahydro,unii-01d3m89a2w,cyclohexene-1,2-dicarboxylic anhydride,3,4,5,6-tetrahydrophthalic acid anhydride,1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione,1, 4,5,6,7-tetrahydro,4,5,6,7-tetrahydro-1,3-isobenzofurandione
12475
4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
C1CCC2=C(C1)C(=O)OC2=O
2426-02-0
C8H8O3
152.15
HMMBJOWWRLZEMI-UHFFFAOYSA-N
3,4,5,6-tetrahydrophthalic anhydride,4,5,6,7-tetrahydroisobenzofuran-1,3-dione,1-cyclohexene-1,2-dicarboxylic anhydride,1,3-isobenzofurandione, 4,5,6,7-tetrahydro,unii-01d3m89a2w,cyclohexene-1,2-dicarboxylic anhydride,3,4,5,6-tetrahydrophthalic acid anhydride,1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione,1, 4,5,6,7-tetrahydro,4,5,6,7-tetrahydro-1,3-isobenzofurandione
12475
4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
C1CCC2=C(C1)C(=O)OC2=O
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Cream
Melting Point 70.0°C to 74.0°C
Quantity 25g
Assay Percent Range 98%
Beilstein 17,461
Solubility Information Solubility in water: reacts.
Formula Weight 152.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 3,4,5,6-Tetrahydrophthalic anhydride
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 219-374-3