Learn More
AdipoGen 3 4 5 6-Tetramethylpyridazin
SDP
Supplier:  AdipoGen CDXT0027M500
Encompass_Preferred
Chemical. CAS 22868-72-0. Formula C8H12N2. MW 136.19. Synthetic. Fluorescent reagent with high selectivity.
Catalog No. 50-256-0046