Learn More
Indofine Chemical 3',4',5,7-Tetrahydr, 90%, 5 Mg
SDP
Supplier:  Indofine Chemical 021342 5 MG
Encompass_Preferred
3',4',5,7-TETRAHYDROXY-3-METHOXYFLAVONE 5 MG, Flavone, Quercetin-3-methylether, 3-O-Methylquercetin, C16H12O7, MW: 316.27, >90% by HPLC, 1486-70-0, MFCD00210588
Catalog No. 50-909-523