Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trichloroaniline 98.0+%, TCI America™
SDP

Catalog No. T30081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30081G Supplier TCI America Supplier No. T30081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 634-91-3
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.46
MDL Number MFCD00007769
InChI Key XOGYQVITULCUGU-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trichloro, ccris 2398, aniline, 3,4,5-trichloro, 3,4,5 trichloroaniline, acmc-1bag0, 3,4,5-trichloro-aniline, 3,4,5-trichlorophenylamine, 3,4,5-trichlorobenzenamine, 3,4,5-tris chloranyl aniline, 3,4,5-trichloroaniline
PubChem CID 12469
IUPAC Name 3,4,5-trichloroaniline
SMILES NC1=CC(Cl)=C(Cl)C(Cl)=C1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 196.46
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3,4,5-Trichloroaniline
EINECSNumber (3)-0290
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.