Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trichloronitrobenzene 96.0+%, TCI America™
SDP

Catalog No. T15805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15805G 5 g
T158025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15805G Supplier TCI America Supplier No. T15805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20098-48-0
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.44
MDL Number MFCD00061096
InChI Key HHLCSFGOTLUREE-UHFFFAOYSA-N
Synonym 3,4,5-trichloronitrobenzene, benzene, 1,2,3-trichloro-5-nitro, 5-nitro-1,2,3-trichlorobenzene, unii-ah0cu8672k, 1,2,3-trichloro-5-nitro-benzene, pubchem3717, acmc-1cmcw, 3,5-trichloronitrobenzene, dsstox_cid_6205, dsstox_rid_78057
PubChem CID 88365
IUPAC Name 1,2,3-trichloro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=C(Cl)C(Cl)=C1

Specifications

Melting Point 71°C
Quantity 5 g
Formula Weight 226.44
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trichloronitrobenzene
EINECSNumber (3)-0459
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.