Learn More
eMolecules​ 14161-11-6 | 3,4,5-Trichloropyridazine | Combi-Blocks | MFCD00006464 | 183.420 | C4HCl3N2 | 98.000 | Clc1cnnc(Cl)c1Cl | 1g | 117561320
Catalog No. 501551246 Shop All eMolecules Inc. Products
Encompass_Preferred
3,4,5-Trichloropyridazine | Combi-Blocks | 14161-11-6 | MFCD00006464 | 183.420 | C4HCl3N2 | 98.000 | Clc1cnnc(Cl)c1Cl | 1g | 117561320
Catalog No. 50-155-1246 Supplier eMolecules​ Supplier No. 117561320
3,4,5-Trichloropyridazine | Combi-Blocks | 14161-11-6 | MFCD00006464 | 183.420 | C4HCl3N2 | 98.000 | Clc1cnnc(Cl)c1Cl | 1g | 117561320