Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trichloropyridine, 98%, Thermo Scientific™

Catalog No. AC458210250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458210250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458210250 Supplier Thermo Scientific Chemicals Supplier No. 458210250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33216-52-3
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.44
InChI Key KKWRVUBDCJQHBZ-UHFFFAOYSA-N
Synonym pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g
PubChem CID 3786531
IUPAC Name 3,4,5-trichloropyridine
SMILES C1=C(C(=C(C=N1)Cl)Cl)Cl

Specifications

Melting Point 72.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 182.44
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.