Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trichloropyridine, 98%

Catalog No. AAA1258703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1258703 1 g
AAA1258706 5 g
AAA1258714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1258703 Supplier Thermo Scientific Chemicals Supplier No. A1258703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 33216-52-3 | C5H2Cl3N | 182.428 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33216-52-3
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.428
MDL Number MFCD00051685
InChI Key KKWRVUBDCJQHBZ-UHFFFAOYSA-N
Synonym pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g
PubChem CID 3786531
IUPAC Name 3,4,5-trichloropyridine
SMILES C1=C(C(=C(C=N1)Cl)Cl)Cl

Specifications

Melting Point 70°C to 74°C
Boiling Point 213°C to 215°C
Quantity 1 g
Beilstein 120542
Formula Weight 182.44
Percent Purity 98%
Chemical Name or Material 3,4,5-Trichloropyridine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.